Learn More
Electron Microscopy Sciences Propylene Oxide, EM Grade, 75-56-9, MFCD00005126, 450ml
Supplier: Electron Microscopy Sciences 20401
Molecular Formula C3H6O, Molecular Weight 58.079, Propylene Oxide, EM Grade is a slovent agent for epoxy resins. It is used widely for infiltration and gradual replacement of the dehydration agent.