Learn More
SMART TUBE INC Programming Station for Smart Tube Base Station
Supplier: SMART TUBE INC PROGRAM1P
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The Programming Station is a hand-held device with an LCD screen and keypad and a simple user interface that is used to author the experimental protocol executed by a Base Station (Models PBS05 and PBS05-M1V-BSM1). The experimental protocol is stored to a Programming Key which resembles a thumb drive. One Programming Key is included. Plugging the Programming Key into a Base Station and powering it on will copy the protocol stored on the Programming Key to the Base Station. User information must be added to all orders for accurate processing
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.