Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProClean™ 2 Scrub Top
Click to view available options
Size:
2X-Large
Description
ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.
Specifications
Size | 2X-Large |
Color | Blue |
Seam Type | Serged |
Sleeve Style | Short Sleeve |
For Use With (Application) | Controlled Environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction