Learn More
Cole-Parmer Oakton SS 2-Cell Conductivity Probe, K=1.0; 2.5 ft Cable
Supplier: Cole-Parmer 3560874
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Conductivity probe, 2-cell, K = 1.0, Ultem body/stainless steel electrode, 2-1/2 ft cable