Learn More
Gilson™ Pro Pipette Service for Single Channel Pipettes
Gilson™ Pro Pipette Service offers quality service, preventative maintenance, and calibration to ensure pipettes lasts for years to come.
$49.00 - $105.00
Specifications
Certifications/Compliance | ISO 17025 Accredited Service |
---|---|
Duration | 5 Days |
For Use With (Equipment) | Single-channel Pipette |
Catalog Number | Mfr. No. | Description | Service Offering | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Service Offering | Product Type | Price | Quantity & Availability | |||
SVC101151G
|
Gilson™
SVC101151 |
Data collected before and after repair | 5 data points collected at 3 volumes | Pipette Pro PM and Calibration Service |
Each for $101.00
|
|
|||
SVC101163G
|
Gilson™
SVC101163 |
Data collected before and after repair | 5 data points collected at 3 volumes | ISO Pipette Pro PM and Calibration Service |
Each for $105.00
|
|
|||
SVC101139G
|
Gilson™
SVC101139 |
Data collected before and after repair | 4 data points collected at 2 volumes | ISO Pipette Pro PM and Calibration Service |
Each for $85.00
|
|
|||
SVC101127G
|
Gilson™
SVC101127 |
Data collected before and after repair | 4 data points collected at 2 volumes | Pipette Pro PM and Calibration Service |
Each for $67.50
|
|
|||
SVC101103G
|
Gilson™
SVC101103 |
Data collected after repair | 2 data points collected at 2 volumes | Pipette Pro PM and Calibration Service |
Each for $49.00
|
|
|||
SVC101115G
|
Gilson™
SVC101115 |
Data collected after repair | 4 data points collected at 2 volumes | Pipette Pro PM and Calibration Service |
Each for $58.25
|
|
|||
Description
- Visual and Functional Inspection
- Internal and External Cleaning
- Leak Test
- Gravimetric Check
- Preventative Maintenance
- Re-grease as needed
- Pipette Label
- Mail-In or Onsite Service Available
- Calibration and Detailed Certificate
- ISO 17025 Calibration Available
- 5 day turn around time
Specifications
ISO 17025 Accredited Service | |
5 Days | |
Single-channel Pipette |