Learn More
Pro-Lab Diagnostics™ AmnioTest™ Kits
Assists in the detection of fetal membrane rupture
$205.00 - $404.00
Specifications
Type | Amniotic Fluid Test Kit |
---|---|
For Use With (Application) | Detection of fetal membrane rupture and also an invaluable tool for OB-GYN and L+D clinics |
Includes | Disposable swabs impregnated with Nitrazine yellow dye packed in individual sterilized sleeve, AmnioTest™ colour card (PL.902) and product instructions for use |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-295-696
|
Pro-Lab Diagnostics™
PL.90120 |
20 Pack |
Pack of 20 for $205.00
|
|
|||||
23-295691
|
Pro-Lab Diagnostics™
PL.901 |
100 Pack |
Pack of 100 for $404.00
|
|
|||||
Description
- Offers an efficient, cost-effective, reliable, and simple-to-use point-of-care screen test to detect amniotic fluid in near-term patients
- An invaluable tool for OB-GYN and L+D clinics
- Sterilized by gamma irradiation
- Packaged individually in peel-pouches, one hundred per box
- Refill buffers and color interpretation cards available, see 22-286-164 and 22-286-158
Specifications
Amniotic Fluid Test Kit | |
Detection of fetal membrane rupture and also an invaluable tool for OB-GYN and L+D clinics | |
Disposable swabs impregnated with Nitrazine yellow dye packed in individual sterilized sleeve, AmnioTest™ colour card (PL.902) and product instructions for use |