Learn More
Novus Biologicals™ PRKRIP1 Overexpression Lysate
Supplier: Novus Biologicals™ NBL114785
Description
The PRKRIP1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
The lysate was created in HEK293T cells, using plasmid ID RC23379 and based on accession number NM_24653. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens PRKR interacting protein 1 (IL11 inducible) (PRKRIP1), mRNA. | |
Native |