Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRKCE Monoclonal Antibody (OTI4G8), TrueMAB™, OriGene
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Mouse Monoclonal Antibody
Supplier: OriGene TA502415S
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Description
PRKCE Monoclonal antibody specifically detects PRKCE in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
PRKCE | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N543 | |
Mouse | |
Affinity chromatography | |
RUO | |
55239 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G8 | |
Unconjugated | |
OGFOD1 | |
5830406C15Rik; MGC125656; MGC125657; nPKC-epsilon; PKC e; PKC[e]; Pkce; Pkcea; PKCepsilon; PRKCE; PRKCH; protein kinase C; protein kinase C epsilon; protein kinase C epsilon type; protein kinase C, epsilon; protein kinase C, eta; R75156 | |
Full length human recombinant protein of human OGFOD1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction