Learn More
Omnimed Inc Economy 3 Section Folding Privacy Screen
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Omnimed Inc 153093
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
*Frame only. Green screens are sold separately as NC1717312* The 3 sections of the screen are attached to each other by a riveted hinge which allows the frame to smoothly fold and unfold as needed. Provides instant isolation for routine, temporary and special circumstances. Anodized aluminum frame and “accordion”hinge allows frame to fold for convenient storage. Screen panels are secured to the frame by top and bottom snap in rods.Features:• Available in 2, 3 or 4 section models• Three sections measures 68”H x 40.5”W• Requires one vinyl or fabric panel per section• Anodized aluminum construction provides a lightweight and durable solution eliminating the possibility of paint chipping or rusting• 5 year warranty
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.