Learn More
Honeywell™ North™ Primair™ 200 Series PA231 Hood Assembly with Fibre-Metal™ E2 Style Hard Hat and Carbtex Cover
Incorporates E2-style hard hat, TyvekTM hood and Carbtex cover for Fibre-Metal welding helmets and faceshields.
Supplier: Honeywell™ PA231E01
Includes: One package of peel-away lens covers.
Description
- Single disposable Tyvek hood features knit neck seal and Quick-On-loop
- Carbtex flame-resistant fabric cover protects user and breathing tube from welding sparks and slag
- E2-style hard hat with ratchet headband, Quick-Lok connections and 8-point suspension fits Fibre-Metal welding helmets and faceshields
- Ratchet adjustment for more secure and comfortable fit
- Manifold directs air over lens to reduce fogging
- For use with North PA700 Series PAPR; and CF1000 Series SAR
Specifications
1000 | |
Continuous Flow | |
One disposable hood with neck seal and Quick-On loop, one flame resistant cover, one E2 hard hat with ratch and Quick-Lok connections for FM welding helmets and faceshields, and one peel-away cover lens | |
PA231E01 |
Loose Fitting Hood | |
Compact Air 200 Series PAPR and CF1000 Series | |
Yes |