Learn More
Cole-Parmer Ashcroft 14902.5 Dual-Scale Low-Pressure Gauge 0 to 3 psi and 0 to 20 kPa
Supplier: Cole-Parmer 6893010
Gauges have brass socket, beryllium copper diaphragm capsule, painted steel case, and polycarbonate lens. Bottom connection is 1/4" NPT(M).