Learn More
Electron Microscopy Sciences Pre-sharpened Double Tip Carbon Rod, 2 3/8 OAL, ¼" diameter
Supplier: Electron Microscopy Sciences 7022102
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2 3/8
Diameter: ¼"
Tip diameter: 0.354"
Tip: 3/16"