Learn More
Electron Microscopy Sciences Pre-sharpened Double-Pointed Carbon Rod, 1½" OAL, ¼" diameter
Supplier: Electron Microscopy Sciences 7022160
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite. Neck type tips.
OAL: 1½"
Diameter: ¼"
Tip diameter: 0.056"
Tip: neck