Learn More
Electron Microscopy Sciences Pre-sharpened Double-Pointed Carbon Rod, 2¼" OAL, ⅛" diameter
Supplier: Electron Microscopy Sciences 7022045
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite. Neck type tips.
OAL: 2¼"
Diameter: ⅛"
Tip diameter: 0.039"
Tip: neck