Learn More
Electron Microscopy Sciences Pre-sharpened Single Tip Carbon Rod, 2-3/8" OAL, ¼" diameter
Supplier: Electron Microscopy Sciences 7022101
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
All of our line of Pre Sharpened Carbon Rods are made from our Spectroscopically Pure Carbon Graphite.
OAL: 2-3/8"
Diameter: ¼"
Tip diameter: 0.205”
Tip: 0.205” neck