Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fischer Technical Company™ Prepared Culture Media in Tubes
Ready-to-use media prepared in disposable borosilicate glass tubes.
Supplier: Fischer Technical Company™ CM901T920
Alert:
Prop 65: Cancer and Reproductive Harm.
Description
- Tubes measure 16 mm O.D. x 125 mm long
- Include black plastic screw caps
- Each tube is labeled with product name, expiration date and lot number
- Refrigerate until ready to use
Order Info
Sold in pack of 20 tubes.
Specifications
Fungi | |
US with Globally Sourced Materials | |
Tubed Media | |
Fungi | |
9 mL | |
20 Tubes/Pk. |
Tubed | |
Sabouraud Dextrose Broth | |
Sterile | |
Microbiological Tests | |
Microbiological Testing |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction