Learn More
Fisherbrand™ Premium Cover Glasses
The ultimate in purity and cleanliness for exacting work in histology, cytology, urinalysis and microbiology
This item is not returnable.
View return policy
Description
- Made of crystal-clear, pure water-white glass with uniform surface quality, precise squared edge, perfect flatness and exact dimensions
- Precleaned and packaged to stay clean
- Each glass is individually inspected and boxed in a molded plastic container for snugness and easy removal
Specifications
Premium Coverslip | |
Glass | |
22 mm | |
30 x 22 mm |
Square | |
30 mm | |
#1 (0.13 to 0.16 mm) | |
1 oz./Pk. |