Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ML Kishigo Premium Black Series™ Surveyors Vest
Click to view available options
Color:
Lime
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Solid polyester front with mesh back
- Heavy duty black bottom to help maintain a clean appearance
- Zipper front closure reinforced with durable webbing
- 2 in. wide high performance reflective material laminated to 3 in. contrasting color
- Left and right mic tabs (Size small only has left mic tab)
- 2 lapel grommets
- Padded neck for comfort
Specifications
Construction | Solid Front, Mesh Back |
Certifications/Compliance | ANSI 107 Class 2 |
Color | Orange |
Size | 5X-Large |
Material | Polyester |
Length (Metric) | 79 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction