Learn More
SCS Premium 3-Layer Vinyl Mat Roll
This three-layer vinyl mat provides excellent electrical properties.
Supplier: SCS TM24600L3GR
Description
- The top layer is made from a tough, easy-to-clean dissipative vinyl
- The center layer, a highly conductive metal scrim, moves the charge from the surface to ground quickly
- The bottom layer is a durable, non-skid, cushion of dissipative foam
- Compatible with constant monitors
- Meets ANSI/ESD S20.20 worksurface required limits and recommendation of ANSI/ESD S4.1
Specifications
50 ft. x 24 in. | |
24 in. | |
60.96 cm | |
15.24 m |
Gray | |
Vinyl | |
0.135 in. | |
50 ft. |