Learn More
MONOPRICE PREM 3D PRINT FILAMENT SILVR
Supplier:  MONOPRICE 12300SILVER
Encompass
Premium 3D Printer Filament PLA 1.75mm 1kg/spool, Silver. 1 kg spool features a thickness of 1.75mm ±0.10mm, with a roundness variation of only 0.03mm. It has a nominal processing temperature of 210°C
Catalog No. NC1758314
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.