Learn More
Labconco Prefilter for Nexus Horizontal Clean Bench
Disposable prefilters trap large particles to extend the HEPA filter life.
Supplier: Labconco 5326800
Description
- MERV 8 rating
- Installation required
- Labconco recommends changing every 3 months, depending on room cleanliness
- Refer to Nexus user guide to determine quantity and amount for your clean bench
Specifications
Enclosures | |
16 x 18 in. | |
Prefilter |