Learn More
Contec™ PREempt™ RTU Disinfectant Solution
Cleans and disinfects environmental surfaces in cleanrooms and laboratory areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces. Contec™ PREempt™ RTU Disinfectant Solution helps to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy.
$75.10 - $351.50
Catalog Number | Mfr. No. | Volume (English) | Container Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Container Type | Price | Quantity & Availability | ||||
19-039-933
|
Contec™
21101 |
Spray bottle |
Case of 12 for $261.50
|
|
|||||
19-039-934
|
Contec™
21105 |
1 gal. | Bottle |
Each for $75.10
Case of 4 EA for $285.50
|
|
||||
19-039-935
|
Contec™
21109 |
5 gal. | Pail |
Each for $351.50
|
|
||||
Description
- AHP - Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase potency and cleaning performance
- Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
- Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
- Responsible - Designed to be easier on surfaces, staff and occupants. Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
- 5 minute tuberculocidal
- 10 minute fungicidal
- 30 seconds broad-spectrum sanitizing
- 1 minute bactericidal, virucidal
Specifications
Disinfectant Solution |