Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contec™ PREempt™ Plus
New and improved formulation of PREempt™ RTU that features faster kill times and lower residue
$130.60 - $149.60
Specifications
Product Type | Cleaner and Disinfectant Solution |
---|---|
pH | 2 to 2.5 |
Sterility | Non-sterile |
Composition | Hydrogen Peroxide |
For Use With (Application) | Cleaning and disinfecting environmental surfaces in cleanrooms and labs |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
19-084-210
|
Contec™
2B101 |
0.9 L |
Case of 12 for $130.60
|
|
|||||
19-084-211
|
Contec™
2B105 |
3.8 L |
Case of 4 for $149.60
|
|
|||||
Description
- Utilizes Accelerated Hydrogen Peroxide™ (AHP™), a patented synergistic blend of commonly used ingredients, when combined with low levels of hydrogen peroxide dramatically increase its efficacy and cleaning performance
- Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and laboratory areas including workstations, fume hoods, equipment and other hard non-porous environmental surfaces
- Ready-to-use, one step broad-spectrum cleaner and disinfectant designed for cleaning and disinfecting hard, non-porous surfaces
- Kills viruses in 30 sec., bacteria in 1 min., fungi in 30 sec. and TB in 1 min.
- Lower residue than original PREempt™ RTU making it more suitable for critical environments
- Contains no volatile organic compounds
- Health hazard rating of zero
Specifications
Cleaner and Disinfectant Solution | |
Non-sterile | |
Cleaning and disinfecting environmental surfaces in cleanrooms and labs |
2 to 2.5 | |
Hydrogen Peroxide |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title