Learn More
MilliporeSigma™ Supelco™ Precolumn Filter Direct-Connect, PEEK
Direct-connect; protects analytical and guard columns.
Supplier: MilliporeSigma™ Supelco™ Z227323
Description
This precolumn filter can be connected directly, hand-tightened, into any HPLC column or guard column that has Valco-compatible end fittings. PEEK cap and body, 2 μm stainless steel frit. For a metal-free system, order PEEK (biocompatible) replacement frits (57430-U).Specifications
PEEK | |
Bore: 0.015 in.; 100°C max. temp.; 5,000 psi max. pressure |