Learn More
World Precision Instrument Precision Inverted Microscope, Trinocular
Supplier: World Precision Instrument PIMT201
Excellent for video recording and photography
The PIMT201 is an affordable inverted microscope for pathologists, biologists and medical researchers. Able to perform intricate and varied applications for vital tissue cultures, it is perfect for observation and education or for professional research in a clinical lab. naturally choose this microscope.
Accessories included: Green, blue and neutral filters, dust cover, immersion oil, manual and warranty card.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.