Learn More
Perkin Elmer US LLC Pre-Assembled Magnetic Steel Screw Cap with PTFE/Silicone (Blue) Septa, 100/pk
Supplier: Perkin Elmer US LLC N6356475
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.