Learn More
Chemglass Life Sciences Pre-Trap Only, 28/15, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF045003
Encompass_Preferred
Pre-Trap Only, 28/15, Airfree, Schlenk. Pre-trap is 51mm OD x 250mm long with two 28/15 socket joints which connect the manifold and main trap assembly which have 28/15 ball joints. Replacement part for AF-0450-01 5-Port Vacuum Manifold. Pre-Tap is labeled G in this picture.
Catalog No. 50-153-0957
Explore available promotions