Learn More
Dynamic Diagnostics Pre-filled 10% Neutral Buffered Formalin (NBF) Specimen Containers
These Pre-filled 10% NBF Specimen Containers are well suited for large throughput laboratories and require a relatively short period of fixation, but can also be used for the long-term storage of tissue. Neutral buffered formalin (NBF) is the most commonly used fixative throughout the world for light microscopy.
$134.00 - $159.00
Specifications
Concentration | 6% Formaldehyde, 4% Alcohol |
---|---|
Grade | Professional Grade |
Product Type | Formalin |
Quantity | 24 Containers/HistoPak Box, 4 HistoPak Boxes/Cs. |
Solution Type | Formalin |
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
22-899-688
|
Dynamic Diagnostics
10150 |
20 mL Total (Pre-filled with 10 mL 10% NBF) |
Case of 96 for $134.00
|
|
|||||
22-899-689
|
Dynamic Diagnostics
10151 |
40 mL Total (Pre-filled with 20 mL 10% NBF) |
Case of 96 for $148.00
|
|
|||||
22-899-690
|
Dynamic Diagnostics
10152 |
60 mL Total (Pre-filled with 40 mL 10% NBF) |
Case of 96 for $159.00
|
|
|||||
22-899-691
|
Dynamic Diagnostics
10153 |
90 mL Total (Pre-filled with 60 mL 10% NBF) |
Case of 96 for $155.00
|
|
|||||
Description
- Pre-filled with 10% neutral buffered formalin
- Packaged in dispenser boxes with perforated tear-away lids to easily access bottles
- Each bottle labeled for clinical use
Specifications
6% Formaldehyde, 4% Alcohol | |
Formalin | |
Formalin |
Professional Grade | |
24 Containers/HistoPak Box, 4 HistoPak Boxes/Cs. | |
Tissue Fixation & Specimen Transport |
Safety and Handling
ShelfLife : Stable under recommended handling and storage conditions
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.