Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Pre-cut Cotton Gauze
SDP

Pre-cut cotton gauze in 4 x 4 in. squares.

Supplier:  StatLab™ 7623

 View more versions of this product

Catalog No. STL7623


Explore available promotions
Add to Cart

Description

Description

  • For general laboratory use or for coverslipping
  • Non-sterile
Specifications

Specifications

Cotton
Non-sterile
10.2 cm
4 in.
Medium
Not Autoclavable
Gauze Pad
4 in.
10.2 cm
200 Bandages/Pk.
Yes
12-ply
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.