Learn More
Biocare Medical, LLC Progesterone Receptor (PR) [PgR636] Antibody - Predilute, 6.0 ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Biocare Medical, LLC APA343AA
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The progesterone receptor is a member of the steroid-receptor family. Steroid hormones bind to intracellular receptors and these receptors can bind to DNA and regulate gene expression directly. Research has shown PR to reflect intact estrogen regulatory machinery and predicts a higher response to endocrine therapy than ER alone. A study has implicated the loss of Progesterone Receptor [PgR636] antibody expression as an independent predictor of poor prognosis and lymph node metastasis in endometrial carcinomas.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.