Learn More
Biotang Inc Accuraci* Disposable Reagent Reservoirs,50ml, 1/pk, 100/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt
Supplier: Biotang Inc 90102
Accuraci* Disposable Reagent Reservoirs,50ml, 1/pk, 100/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt