Learn More
Cytiva Polypropylene/Polyethylene Separator Depth Filters - 10 m, 47 mm (100/pkg)
Supplier: Cytiva 60344
Virtually Inert to Chemical and Biological Agents-Outstanding wet strength-High chemical resistance-Provides a channeling effect for liquid, increasing overall filter flow rate when used in series with submicron membrane filters-Ensures stability when filtering concentrated acids, alkalis, or oxidizers This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.