Learn More
Andwin Scientific POWER TROL 120V
SDP
Supplier:  Andwin Scientific 104APL120
Encompass_Preferred
The 120V Glas-Col PowrTrol Proportional Voltage Control is a compact device that is ideal for the regulation of the input voltage of resistive devices by simple turning of the dial knob. As the number on the ideal increases, the hotter the load will be. This product is sold with a 6 feet attached power cord and grid support bracket, which allows fume hood mounting for the maximization of bench space.
Catalog No. NC9587142