Learn More
Includes: Intruder Kit Contents:
- P-16 Rescue Tool (1 each)
- Attachment Pin Set (1 each)
- Power Cable (1 each)
- Controller Unit (1 each)
- 12VDC Power Pack (1 each)
- Jumper Cables (1 each)
- Battery Charger (1 each)
- Spreader Arm Set (1 each)
- Curved Blade Cutter (1 each)
- Straight-blade Cutter (1 each)
Description
Utilizing aerospace technology developed for military fighter jet control applications, this all-electric, “jaws-type” system operates completely without hydraulics for exceptional versatility and power. Highly effective for use in numerous rescue applications including, vehicle extrication, collapse and remote rescues, indoor and confined-space operations, forcible entry, and special operations.
- Lightweight and compact for portability and ease of use
- No set-up necessary for rapid response
- Safe for indoor/outdoor use—runs in O2 depleted atmosphere
- Interchangeable spreaders and cutters maximize versatility and minimize compartment space
- Simultaneously operates accessories such as Milwaukee™ Sawzall™, Ramfan™ blowers, lights, and more with 12VDC power pack
- Ambidextrous controls; color-coded for enhanced safety and ease of operation
- Patented 70° swivel-head for around-the-corner purchase points
- Operates quietly and requires little maintenance
Power Hawk Rescue System components and accessories available in various kit configurations and individually.
Certifications
P-16 Rescue Tool (19-500-392), Power Pack (NC9164139), and Power Pusher Rams (19-817-201 and 19-500-393) are UL certified, compliant to NFPA 1936.
Specifications
In-1 Intruder Kit | |
P-16 rescue tool, AP-1600 attachment pins, CA-4M power cable, PC-100 controller unit, PWR-3X12 power pack, BC-U1 battery charger, S-1601 spreader arm, C-1601 curved-blade cutter, CS-1602 straight blade cutter, JC4-16 jumper cables, 16 ft. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.