Learn More
Cole-Parmer Digi-Sense 104A PL312 Variable- Time Output Controller 1440 Watts, 120 V 50/60 Hz
Supplier: Cole-Parmer 3622591
This timed control turns the output load on and off and then repeats the cycle. Each position on the knob from LO to HI represents a specific cycle time where the on time ranges from 6 to 100 percent. For example the LO setting represents a total cycle of 60 seconds with an on time of 4 seconds. This type of control can be used to regulate the temperature of resistive loads. The pilot light on the front will glow when the load is on.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.