Learn More
Eisco™ Plastic Powder Funnels
Eisco™ Plastic Powder Funnels are an ideal method for transferring powder reagents from one vessel to another.
Supplier: Eisco™ CH0457B
Description
- Polypropylene powder funnel
- Designed for transferring powder reagents
- Translucent
- Durable for student use
- Easy to clean
- Autoclavable
- Chemical, acid, and alkali resistant
Specifications
80 mm | |
Yes | |
Parallel stem |
Polypropylene | |
Transferring powdered reagents | |
Powder Funnel |