Learn More
Microsolv Technology Corporation Pouring Ring, Replacement, Natural, for HPLC Mobile Phase Reservoir Bottles. GL45 Threads. 1 EA.
Supplier: Microsolv Technology Corporation 5880745
Using this Natural color Polypropylene Pouring or "Drip Ring" can make decanting liquids from any bottle with a GL45 Thread easier and prevent that last drop from getting away from you. This ring will fit deeper into the mouth of the Bottle, making is more secure and less likely to fall out.
This item is not returnable.
View return policy