Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Sodium Tartrate, Tetrahydrate, Powder, USP, 99-102%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Potassium Sodium Tartrate, Tetrahydrate, Powder, USP is sometimes referred to as Rochelle or Seignette's salt and used as a laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Packaging | Fiber Drum |
Quantity | 50 kg |
Percent Purity | 99 to 102% |
Grade | USP |