Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Permanganate, Certified, 0.200N ±0.001N (0.040M), LabChem™
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Specifications
Color | Purple |
CAS Max % | 99.37, 0.63 |
Physical Form | Liquid |
Quantity | 10 L |
Linear Formula | KMnO4 |
Solubility Information | Soluble in water |
Formula Weight | 158.04 |
Grade | Certified |
Concentration or Composition (by Analyte or Components) | 0.200N ±0.001N (0.040M) |
Identification | Passes Test |
Show More |
Safety and Handling
Hazard Statement | GHS H Statement Harmful to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Avoid release to the environment. Dispose of contents/container to comply with local, state and federal regulations. |
Recommended Storage | Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction