Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 0.5 N, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88708
Click to view available options
Quantity:
500 mL

Chemical Identifiers

CAS 7722-64-7, 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.03
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
IUPAC Name potassium manganesoylolate
SMILES [K+].[O-][Mn](=O)(=O)=O

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.5 N
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.