Learn More
MEDTOX™ Positive Control for Profile-V MEDTOXScan™ test systems
Monitors the performance of Profile-V MEDTOXScan™ drugs of abuse test devices. MEDTOX™ Positive Controls for Profile-V MEDTOXScan test systems are compatible with all quantitative and qualitative drug detection procedures that are sufficiently sensitive to detect the control constituents.
Specifications
Sample Type Urine
Intended Use Positive control used for quality control with MEDTOXScan drugs of abuse tests
Packaging Bottle
Quantity 5 mL
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-000-403
View Documents
MEDTOX™
102367
OPI100
Each for $41.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-000-404
View Documents
MEDTOX™
102365
OPI2000
Each for $45.75
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Positive control urines have been spiked with authentic reference drug standards and/or appropriate metabolites
  • Should be treated as any “unknown” specimen while following the specific protocol of the assay being used
  • Controls prepared from human based urine available as a negative and Cutoff +50% to monitor the performance of MEDTOX drugs of abuse test devices
  • Each bottle contains stabilized human based urine
Specifications
Specifications
Urine
Positive control used for quality control with MEDTOXScan drugs of abuse tests
Bottle
5 mL