Learn More
Electron Microscopy Sciences Porta-Vac II with Peek Tip for 6" wafers, Molded PEEK wafer tip and 9V Disposable Battery
SDP
Supplier:  Electron Microscopy Sciences 7192901
Encompass_Preferred
The Porta-Vac II wafer handling toolkit comes complete with a disposable 9V battery and a wafer tip. A large switch provides easy on/off control. The internal air filter is accessable from the front of the tool, and an internal exhaust filter ensures better than Class 1 performance.
Includes: Molded PEEK wafer tip and 9V Disposable Battery
Catalog No. 50-193-1148