Learn More
TSI Incorporated PortaCount Respirator Fit Tester
Supplier: TSI Incorporated 8048T
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The new PortaCount Respirator Fit Tester is a quantitative respirator fit tester. It features intelligent touchscreen solutions that support respiratory protection programs from training through compliance. The 8048-T Kit lets you begin testing immediately, with everything you need for accurate, easy, fast, and flexible respirator fit testing sessions.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.