Learn More
Scientific Gear LLC Portable Density/Specific Gravity Meter
SDP
Supplier:  Scientific Gear LLC DA130N
Encompass_Preferred
The DA-130N portable density/specific gravity meter has 12 measurement modes including density of alcohol, API, Baume, Brix, Plato, Proof, H2S04, temp. compensated density, temp. compensated specific gravity, concentrations and more.
Catalog No. NC2085372