Learn More
Broadley James Corporation Porous Sparger, 5 micron
Supplier: Broadley James Corporation B422202005
Attaches to the tip of the Sparger Tube, this 316L stainless steel sintered tip is available in a variety of pore sizes.