Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ POP-6™ Polymer for the 310 Genetic Analyzer
POP-6™ is a denaturing polymer used on the 310 genetic analyzer. POP-6™ Polymer is used for sequencing applications.
Supplier: Applied Biosystems™ 402837
Description
- Easy to Use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
- Robust Formulation: Formulated for sequencing and fragment analysis applications
- Replenishable: Allow capillaries to be used multiple times
Specifications
98.5% up to 400 bases (for 36 cm capillary), 98.5% up to 600 bases (for 50 cm capillary) | |
310 Genetic Analyzer | |
Stability on instrument 7 days, Up to 250 samples/3.5 ml | |
DNA Sequencing Reagents | |
1 hr for 36 cm capillary, 2 hrs 45 min for 50 cm capillary |
Store at 2°C to 8°C. | |
Standard Sequencing, Rapid Sequencing | |
POP-6™ | |
3 mL |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction