Learn More
Innovative Research Inc Pooled Human Plasma (Blood Derived) Na Citrate 50ml
Supplier:  Innovative Research Inc IPLAWBNAC50ML
Encompass_Preferred
Pooled Human Plasma (Blood Derived) from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center and processed into plasma via cetrifugation. The plasma is frozen immediately after processing. Please note that this product does not allow for donor requests off-the-shelf, but we can create custom pooled lots to your exact specifications upon request. Contact us for details & pricing.
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. If you would like any further testing or analysis performed prior to shipping, please let us know. 
Anticoagulants
We are able to collect this product using the anticoagulant that will work best for your research. Our standard anticoagulant options include K2 EDTA, K3 EDTA, Na Citrate, and Li Heparin, with o
Catalog No. 50-203-6380
Due to product restrictions, please to purchase or view availability for this product.