Learn More
Kinematica™ POLYTRON™ PT 2500E Bundle with Generator
The sleek, sound-insulated casing with a digital speed display conceals enough power to handle a variety of applications safely and quickly. Our broad range of dispersing aggregates cover the range from preparing samples to testing product formulas and recipes.
Supplier:  Kinematica™ 11070012
 View more versions of this product
Catalog No. 08-451-058
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Variable speed control with digital display
  • High-speed, high-performance motor
  • Soft-start motor prevents splashing and sample loss
  • Quick-connect: change generator with one hand
  • Accept wide range of stainless-steel Polytron generators
Specifications
Specifications
30,000 rpm
50 Hz
0.1 to 5 mL
Includes 5 mm generator
110/115 V
5 mL
Kinematica Polytron
Product Suggestions
Product Suggestions