Learn More
Hach Company BOD Seed Inoculum, Polyseed, pk/50 capsules
Supplier: Hach Company 2918700
BOD Seed Inoculum for use in Biochemical Oxygen Demand (BOD5) test procedures. A blend of specialized microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses. Package of 50 capsules. International Laboratory Supply (InterLab), LTD* Consitent seed source: same bacteria, same type of bacteria, same amount of bacteria blend* Known bacterial content is free from pathogens* Save and convenient pre-measured dry product in easy to use capsules* Two years shelf life
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.