Learn More
Fisherbrand™ Polypropylene Tubing Equal Angle Y-Shaped Connectors
Highly resistant to many chemicals.
Description
- Excellent chemical resistance
- Autoclavable polypropylene
- Serrated connecting surfaces to prevent tubing slipping
- Withstands flows to 20 p.s.i.
- The elasticity of the tubing will influence choice of size
- Dimensions listed are nominal
Specifications
Equal Angle Tubing Connector | |
0.25 to 0.375 in. | |
2.23 in. | |
6.4 to 9.5 mm | |
0.4 in. | |
43.8 mm | |
White |
Autoclavable | |
Polypropylene | |
56.6 mm | |
10.16 mm | |
Y | |
1.7 in. | |
20 psi |